accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6975-91-3

IUPAC Name: cyclohexyl 2-chloroacetate
Synonyms: cyclohexyl 2-chloroacetate
Molecular Weight (g/mol): 176.64
Molecular Formula: C8H13ClO2
InChi Key: IBFHBLOKQVCABG-UHFFFAOYSA-N
SMILES: ClCC(=O)OC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results