accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HONOHOHOHHO

CAS RN 6976-37-0

IUPAC Name: 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Synonyms: bis-tris buffer bis-tris methane bis-hydroxyethyl tromethamine bistris bis-tris
Molecular Weight (g/mol): 209.24
Molecular Formula: C8H19NO5
InChi Key: OWMVSZAMULFTJU-UHFFFAOYSA-N
SMILES: OCCN(CCO)C(CO)(CO)CO
Quantity 
  • (4)
  • (1)
  • (10)
  • (4)
  • (2)
  • (4)
  • (4)
  • (1)
  • (8)
Molecular Weight (g/mol) 
  • (11)
  • (23)
Physical Form 
  • (9)
  • (3)
  • (11)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (6)
  • (1)
  • (7)
  • (3)
  • (5)
pH 
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (7)
  • (3)
  • (8)
  • (11)
  • (3)
  • (3)
  • (4)
  • (1)
  • (10)
  • (4)
  • (2)
  • (4)
  • (4)
  • (1)
  • (8)
  • (11)
  • (23)
  • (9)
  • (3)
  • (11)
  • (2)
  • (2)
  • (2)
  • (6)
  • (1)
  • (7)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (8)
  • (3)
  • (2)
  • (2)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)