accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 69762-88-5

IUPAC Name: tributyl[6-(tributylazaniumyl)hexyl]azanium dihydroxide
Synonyms: tributyl[6-(tributylammonio)hexyl]azanium dihydroxide
Molecular Weight (g/mol): 488.89
Molecular Formula: C30H68N2O2
InChi Key: ALYGOOWRCDZDTJ-UHFFFAOYSA-L
SMILES: [OH-].[OH-].CCCC[N+](CCCC)(CCCC)CCCCCC[N+](CCCC)(CCCC)CCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results