accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 69778-83-2

IUPAC Name: 4-methoxy-2,5-dihydro-1H-pyrrol-2-one
Synonyms: 4-methoxy-1,5-dihydropyrrol-2-one
Molecular Weight (g/mol): 113.12
Molecular Formula: C5H7NO2
InChi Key: TXKQBYYDTLOLHA-UHFFFAOYSA-N
SMILES: COC1=CC(=O)NC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results