accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3PH3CCH3CH3PH3CCH3Pd

CAS RN 69861-71-8

IUPAC Name: bis(tris(2-methylphenyl)phosphane) palladium
Synonyms: bis(tris(2-methylphenyl)phosphane) palladium
Molecular Weight (g/mol): 715.17
Molecular Formula: C42H42P2Pd
InChi Key: CUBIJGNGGJBNOC-UHFFFAOYSA-N
SMILES: [Pd].CC1=CC=CC=C1P(C1=CC=CC=C1C)C1=CC=CC=C1C.CC1=CC=CC=C1P(C1=CC=CC=C1C)C1=CC=CC=C1C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results