CAS RN 6989-24-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bayogenin, TRC
CAS: 6989-24-8 Molecular Formula: C30H48O5 Molecular Weight (g/mol): 488.7 Synonym: Olean-12-en-28-oic acid, 2,3,23-trihydroxy-, (2β,3β,4α)-,Olean-12-en-28-oic acid, 2β,3β,23-trihydroxy- (7CI,8CI),2β,3β,23α-Trihydroxy-12-oleanen-28-oic acid,Bayogenin,Bayogenine IUPAC Name: (4aS,6aR,6aS,6bR,8aR,9R,10R,11S,12aR,14bS)-10,11-dihydroxy-9-(hydroxymethyl)-2,2,6a,6b,9,12a-hexamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid SMILES: CC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@H](O)[C@H](O)[C@@](C)(CO)[C@@H]5CC[C@@]34C)[C@@H]2C1)C(=O)O