accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

A(a)

CAS RN 69912-79-4

IUPAC Name: aluminium(3+) sodium bis(oxosilanebis(olate))
Synonyms: sodium aluminosilicate sodium silicoaluminate
Molecular Weight (g/mol): 202.14
Molecular Formula: AlNaO6Si2
InChi Key: URGAHOPLAPQHLN-UHFFFAOYSA-N
SMILES: [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
Quantity 
  • (6)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
Pore Size 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (7)
  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (2)