accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 70-53-1

IUPAC Name: hydrogen 2,6-diaminohexanoic acid chloride
Synonyms: hydrogen lysine chloride
Molecular Weight (g/mol): 182.65
Molecular Formula: C6H15ClN2O2
InChi Key: BVHLGVCQOALMSV-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NCCCCC(N)C(O)=O
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)