accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 700-00-5

IUPAC Name: 9-methyl-9H-purin-6-amine
Synonyms: 9H-purin-6-amine, 9-methyl-
Molecular Weight (g/mol): 149.16
Molecular Formula: C6H7N5
InChi Key: WRXCXOUDSPTXNX-UHFFFAOYSA-N
SMILES: CN1C=NC2=C(N)N=CN=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)