CAS RN 700-12-9
IUPAC Name:
1,2,3,4,5-pentamethylbenzene
Synonyms:
pentamethylbenzene pentamethylbenzol
Molecular Weight (g/mol):
148.25
Molecular Formula:
C11H16
InChi Key:
BEZDDPMMPIDMGJ-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C(C)C(C)=C1C
1
–
1
of
1
results
1
Pentamethylbenzene, 99%
CAS: 700-12-9 Molecular Formula: C11H16 Molecular Weight (g/mol): 148.249 MDL Number: MFCD00008522 InChI Key: BEZDDPMMPIDMGJ-UHFFFAOYSA-N Synonym: pentamethylbenzene,benzene, pentamethyl,pentamethylbenzol,benzene, 1,2,3,4,5-pentamethyl,pentamethyl phenyl,acmc-1bkt8,ksc492e3d,1,3,4,5-pentamethylbenzene,pentamethylbenzene 5g PubChem CID: 12784 ChEBI: CHEBI:38998 IUPAC Name: 1,2,3,4,5-pentamethylbenzene SMILES: CC1=CC(=C(C(=C1C)C)C)C