accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 700-13-0

IUPAC Name: 2,3,5-trimethylbenzene-1,4-diol
Synonyms: 2,3,5-trimethylbenzene-1,4-diol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: AUFZRCJENRSRLY-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C(C)C(C)=C1O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results