accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 700-49-2

IUPAC Name: 2-fluoro-7H-purin-6-amine
Synonyms: 2-fluoro-7H-purin-6-amine
Molecular Weight (g/mol): 153.12
Molecular Formula: C5H4FN5
InChi Key: WKMPTBDYDNUJLF-UHFFFAOYSA-N
SMILES: NC1=C2NC=NC2=NC(F)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results