CAS RN 700-82-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Hexahydrocoumarin, TRC
CAS: 700-82-3 Molecular Formula: C9 H12 O2 Molecular Weight (g/mol): 152.19 Synonym: Coumarin, 3,4,5,6,7,8-hexahydro- (6CI,7CI,8CI),3,4,5,6,7,8-Hexahydro-2H-1-benzopyran-2-one,1-Cyclohexene-1-propanoic acid, 2-hydroxy-, δ-lactone,3,4,5,6,7,8-Hexahydrocoumarin,Hexahydrocoumarin,δ-1,6-2-Oxabicyclo(4.4.0)decen-3-one IUPAC Name: 3,4,5,6,7,8-hexahydrochromen-2-one SMILES: O=C1CCC2=C(CCCC2)O1
2
3,4,5,6,7,8-Hexahydrocoumarin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality