accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7003-32-9

CH3H2N

CAS RN 7003-32-9

IUPAC Name: 2-methylcyclohexan-1-amine
Synonyms: 2-methylcyclohexan-1-amine
Molecular Weight (g/mol): 113.2
Molecular Formula: C7H15N
InChi Key: FEUISMYEFPANSS-UHFFFAOYNA-N
SMILES: CC1CCCCC1N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results