CAS RN 7008-26-6
CAS RN 7008-26-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dichlorisone, TRC
CAS: 7008-26-6 Molecular Formula: C21 H26 Cl2 O4 Molecular Weight (g/mol): 413.33 Synonym: (11β)-9,11-Dichloro-17,21-dihydroxypregna-1,4-diene-3,20-dione,9,11β-Dichloro-17,21-dihydroxypregna-1,4-diene-3,20-dione,9α,11β-Dichloro-17α,21-dihydroxypregna-1,4-diene-3,20-dione; IUPAC Name: (8S,9R,10S,13S,14S,17R)-9,11-dichloro-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one SMILES: C[C@]12CC(Cl)[C@@]3(Cl)[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO