accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SOOClF

CAS RN 701-27-9

IUPAC Name: 3-fluorobenzene-1-sulfonyl chloride
Synonyms: 3-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 194.6
Molecular Formula: C6H4ClFO2S
InChi Key: OKYSUJVCDXZGKE-UHFFFAOYSA-N
SMILES: FC1=CC(=CC=C1)S(Cl)(=O)=O
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)