accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NOFCl

CAS RN 701-43-9

IUPAC Name: 3-chloro-4-fluorobenzamide
Synonyms: 3-chloro-4-fluorobenzamide
Molecular Weight (g/mol): 173.57
Molecular Formula: C7H5ClFNO
InChi Key: AEYMQEMBMKCNEG-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(F)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results