accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 70131-67-8

IUPAC Name: hydroxy-λ¹-silanylidyne
Synonyms: hydroxy-λ¹-silanylidyne
Molecular Weight (g/mol): 45.09
Molecular Formula: HOSi
InChi Key: XQSFXFQDJCDXDT-UHFFFAOYSA-N
SMILES: O[Si]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results