accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HNOCH3

CAS RN 702-24-9

IUPAC Name: [(4-methoxyphenyl)methyl](methyl)amine
Synonyms: [(4-methoxyphenyl)methyl](methyl)amine
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: AIJFPNKGGAPZFJ-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(OC)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (5)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)
  • (2)