accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3

CAS RN 702-79-4

IUPAC Name: 1,3-dimethyladamantane
Synonyms: 1,3-dimethyladamantane
Molecular Weight (g/mol): 164.29
Molecular Formula: C12H20
InChi Key: CWNOIUTVJRWADX-UHFFFAOYSA-N
SMILES: CC12CC3CC(C1)CC(C)(C3)C2
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results