accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

MgBr

CAS RN 703-55-9

IUPAC Name: magnesium(2+) naphthalen-1-ide bromide
Synonyms: magnesium(2+) naphthalen-1-ide bromide
Molecular Weight (g/mol): 231.38
Molecular Formula: C10H7BrMg
InChi Key: VCBWOINKHDKRAF-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].C1=CC=C2[C-]=CC=CC2=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results