accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOO

CAS RN 703-59-3

IUPAC Name: 3,4-dihydro-1H-2-benzopyran-1,3-dione
Synonyms: 4H-2-benzopyran-1,3-dione
Molecular Weight (g/mol): 162.14
Molecular Formula: C9H6O3
InChi Key: AKHSBAVQPIRVAG-UHFFFAOYSA-N
SMILES: O=C1CC2=CC=CC=C2C(=O)O1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)