accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 70374-39-9

IUPAC Name: (3E)-6-chloro-3-{hydroxy[(pyridin-2-yl)amino]methylidene}-2-methyl-2H,3H,4H-1λ⁶-thieno[2,3-e][1,2]thiazine-1,1,4-trione
Synonyms: telos lorcam xafon lorcam (TN) XEFO taigalor acabel safem xefocam lornoxicamum
Molecular Weight (g/mol): 371.81
Molecular Formula: C13H10ClN3O4S2
InChi Key: OXROWJKCGCOJDO-JLHYYAGUSA-N
SMILES: CN1\C(=C(\O)NC2=CC=CC=N2)C(=O)C2=C(C=C(Cl)S2)S1(=O)=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)