accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 704-04-1

H3COFONH2

CAS RN 704-04-1

IUPAC Name: 5-fluoro-2-methoxybenzamide
Synonyms: 5-fluoro-2-methoxybenzamide
Molecular Weight (g/mol): 169.16
Molecular Formula: C8H8FNO2
InChi Key: CJDKHPOEWZBRAJ-UHFFFAOYSA-N
SMILES: COC1=C(C=C(F)C=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results