CAS RN 7045-71-8
CAS RN 7045-71-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Methylundecane, TRC
CAS: 7045-71-8 Molecular Formula: C12H26 Molecular Weight (g/mol): 170.33 IUPAC Name: 2-methylundecane SMILES: CCCCCCCCCC(C)C