accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNClFFFCl

CAS RN 705-24-8

IUPAC Name: 4,6-dichloro-2-(trifluoromethyl)pyrimidine
Synonyms: 4,6-dichloro-2-(trifluoromethyl)pyrimidine
Molecular Weight (g/mol): 216.97
Molecular Formula: C5HCl2F3N2
InChi Key: QFWVAJQVYBRTCL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC(Cl)=CC(Cl)=N1
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)