accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFCl

CAS RN 705-29-3

IUPAC Name: 1-(chloromethyl)-3-(trifluoromethyl)benzene
Synonyms: 1-(chloromethyl)-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 194.58
Molecular Formula: C8H6ClF3
InChi Key: XGASTRVQNVVYIZ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(CCl)=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)