accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7057-57-0

IUPAC Name: zinc(2+) 8-(dimethylamino)-10λ⁴-oxa-5-azatetraphen-10-ylium trichloride
Synonyms: zinc(2+) 8-(dimethylamino)-10λ⁴-oxa-5-azatetraphen-10-ylium trichloride
Molecular Weight (g/mol): 447.06
Molecular Formula: C18H15Cl3N2OZn
InChi Key: YTEJSAFVYHDCSN-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C=C3

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results