accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 706806-60-2

IUPAC Name: (2R)-2-benzyl-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid
Synonyms: (2R)-2-benzyl-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 305.37
Molecular Formula: C17H23NO4
InChi Key: JUUNPRGYFCIXSM-QGZVFWFLSA-N
SMILES: CC(C)(C)OC(=O)N1CCC[C@@]1(CC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results