accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 70693-62-8

IUPAC Name: pentapotassium (hydroperoxysulfonyl)oxidanide oxidanidesulfonoperoxoate dihydrogen sulfate
Synonyms: pentapotassium (hydroperoxysulfonyl)oxidanide peroxymonosulfate dihydrogen sulfate
Molecular Weight (g/mol): 614.74
Molecular Formula: H3K5O18S4
InChi Key: HJKYXKSLRZKNSI-UHFFFAOYSA-I
SMILES: [K+].[K+].[K+].[K+].[K+].OS([O-])(=O)=O.OS([O-])(=O)=O.OOS([O-])(=O)=O.[O-]OS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Grade 
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)