accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 70764-39-5

OHOF

CAS RN 70764-39-5

IUPAC Name: [2-(4-fluorophenoxy)phenyl]methanol
Synonyms: [2-(4-fluorophenoxy)phenyl]methanol
Molecular Weight (g/mol): 218.23
Molecular Formula: C13H11FO2
InChi Key: LHUOIWVHXSPAQX-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1OC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results