accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7080-50-4

CH3SOONClNaH2OH2OH2O

CAS RN 7080-50-4

IUPAC Name: (chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate
Synonyms: (chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate
Molecular Weight (g/mol): 204.65
Molecular Formula: C7H7ClNO2S
InChi Key: VQWVWLDBJISZHX-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1)S([O-])(=O)=NCl
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (4)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (4)
  • (2)
Color 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)