accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 709-09-1

H3COOCH3NOO

CAS RN 709-09-1

IUPAC Name: 1,2-dimethoxy-4-nitrobenzene
Synonyms: 1,2-dimethoxy-4-nitrobenzene
Molecular Weight (g/mol): 183.16
Molecular Formula: C8H9NO4
InChi Key: YFWBUVZWCBFSQN-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)[N+]([O-])=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results