accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOINOO

CAS RN 709-49-9

IUPAC Name: 1-iodo-2,4-dinitrobenzene
Synonyms: 1-iodo-2,4-dinitrobenzene 4-iodo-1,3-dintrobenzene 6-iodo-1,3-dintrobenzene
Molecular Weight (g/mol): 294
Molecular Formula: C6H3IN2O4
InChi Key: FXMKXMJLXRTQSW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(I)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results