CAS RN 71-62-5
IUPAC Name:
(1R,2S,6S,9S,10R,11S,12S,14R,15S,18S,19S,22S,23S,25R)-1,10,11,12,14,23-hexahydroxy-6,10,19-trimethyl-24-oxa-4-azaheptacyclo[12.12.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁵.0¹⁸,²³.0¹⁹,²⁵]hexacosan-22-yl 3,4-dimethoxybenzoate
Synonyms:
veratridine veratrine (amorphous) 3-veratroylveracevine veratridin veratridine (substance) veratrine
Molecular Weight (g/mol):
673.8
Molecular Formula:
C36H51NO11
InChi Key:
FVECELJHCSPHKY-YFUMOZOISA-N
SMILES:
COC1=CC=C(C=C1OC)C(=O)O[C@H]1CC[C@@]2(C)[C@@H]3CC[C@H]4[C@]5(O)C[C@H](O)[C@@]6(O)[C@@H](CN7C[C@@H](C)CC[C@H]7[C@@]6(C)O)[C@]5(O)C[C@]24O[C@]13O
1
–
3
of
3
results
1
R&D Systems™ Veratridine
CAS: 71-62-5 Molecular Formula: C36H51NO11
2
Veratridine, TRC
CAS: 71-62-5 Molecular Formula: C36 H51 N O11 Molecular Weight (g/mol): 673.79 Synonym: Cevane-3,4,12,14,16,17,20-heptol, 4,9-epoxy-, 3-(3,4-dimethoxybenzoate), (3β,4α,16β)-,Veratridine (6CI,7CI,8CI),3-Veratroylveracevine,NSC 7524,Veracevine, veratrate SMILES: COc1ccc(cc1OC)C(=O)O[C@H]2CC[C@@]3(C)[C@@H]4CC[C@H]5[C@]6(O)C[C@H](O)[C@@]7(O)[C@@H](CN8C[C@@H](C)CC[C@H]8[C@@]7(C)O)[C@]6(O)C[C@]35O[C@]24O
3
Thermo Scientific™ Veratridine, ≥97%
Activates sodium ion channels