accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7103-09-5

CH2MgBr

CAS RN 7103-09-5

IUPAC Name: magnesium(2+) but-3-en-1-ide bromide
Synonyms: magnesium(2+) but-3-en-1-ide bromide
Molecular Weight (g/mol): 159.31
Molecular Formula: C4H7BrMg
InChi Key: JAUWOQLHLFMTON-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].[CH2-]CC=C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)