CAS RN 711-01-3
IUPAC Name:
methyl adamantane-1-carboxylate
Synonyms:
methyl adamantane-1-carboxylate
Molecular Weight (g/mol):
194.27
Molecular Formula:
C12H18O2
InChi Key:
CLYOOVNORYNXMD-UHFFFAOYSA-N
SMILES:
COC(=O)C12CC3CC(CC(C3)C1)C2
1
–
3
of
3
results
1
1-Adamantanecarboxylic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Adamantanecarboxylic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.