accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOSN

CAS RN 7113-10-2

IUPAC Name: 2-phenyl-1,3-thiazole-4-carboxylate
Synonyms: 2-phenyl-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol): 204.22
Molecular Formula: C10H6NO2S
InChi Key: IBUSLNJQKLZPNR-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CSC(=N1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results