accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7120-43-6

H2NOClHO

CAS RN 7120-43-6

IUPAC Name: 5-chloro-2-hydroxybenzamide
Synonyms: 5-chloro-2-hydroxybenzamide
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: NNHMQZBVJPQCAK-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(Cl)=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results