accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 71219-80-2

ONCO

CAS RN 71219-80-2

IUPAC Name: 1-isocyanato-3-(phenoxymethyl)benzene
Synonyms: 1-isocyanato-3-(phenoxymethyl)benzene
Molecular Weight (g/mol): 225.25
Molecular Formula: C14H11NO2
InChi Key: CXYJFMVKSSTBDJ-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=CC(COC2=CC=CC=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results