accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7128-64-5

IUPAC Name: 5-tert-butyl-2-[5-(5-tert-butyl-1,3-benzoxazol-2-yl)thiophen-2-yl]-1,3-benzoxazole
Synonyms: bisthiophene
Molecular Weight (g/mol): 430.57
Molecular Formula: C26H26N2O2S
InChi Key: AIXZBGVLNVRQSS-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C2OC(=NC2=C1)C1=CC=C(S1)C1=NC2=CC(=CC=C2O1)C(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)