accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 713-52-0

IUPAC Name: 5-(methoxycarbonyl)-2-nitrobenzen-1-olate
Synonyms: 5-(methoxycarbonyl)-2-nitrobenzenolate
Molecular Weight (g/mol): 196.14
Molecular Formula: C8H6NO5
InChi Key: UEGCRFNWTGYVKX-UHFFFAOYSA-M
SMILES: COC(=O)C1=CC=C(C([O-])=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results