accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7138-15-0

IUPAC Name: 3-bromo-2-nitroaniline
Synonyms: 3-bromo-2-nitroaniline
Molecular Weight (g/mol): 217.02
Molecular Formula: C6H5BrN2O2
InChi Key: GLKHLBAXHLAQBJ-UHFFFAOYSA-N
SMILES: NC1=C(C(Br)=CC=C1)[N+]([O-])=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results