accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFOHNOHNClONClFFFCl

CAS RN 71422-67-8

IUPAC Name: 1-(3,5-dichloro-4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}phenyl)-3-(2,6-difluorobenzoyl)urea
Synonyms: chlorfluazuron atabron
Molecular Weight (g/mol): 540.65
Molecular Formula: C20H9Cl3F5N3O3
InChi Key: UISUNVFOGSJSKD-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1C(=O)NC(=O)NC1=CC(Cl)=C(OC2=NC=C(C=C2Cl)C(F)(F)F)C(Cl)=C1
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results