accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Cl

CAS RN 71434-47-4

IUPAC Name: (7-chloroheptyl)benzene
Synonyms: (7-chloroheptyl)benzene
Molecular Weight (g/mol): 210.75
Molecular Formula: C13H19Cl
InChi Key: PNUBJDFTMKBZJK-UHFFFAOYSA-N
SMILES: ClCCCCCCCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)