accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7149-69-1

IUPAC Name: 1,3-dichloro-2-methyl-5-nitrobenzene
Synonyms: 1,3-dichloro-2-methyl-5-nitrobenzene
Molecular Weight (g/mol): 206.02
Molecular Formula: C7H5Cl2NO2
InChi Key: RUVCGWLHTVGNGI-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=C(C=C1Cl)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results