accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COH3COHO

CAS RN 7151-68-0

IUPAC Name: 3-methoxy-4-methylbenzoic acid
Synonyms: 3-methoxy-4-methylbenzoic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: CEAVPXDEPGAVDA-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1C)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results