accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOBr

CAS RN 7154-66-7

IUPAC Name: 2-bromobenzoyl chloride
Synonyms: 2-bromo-benzoyl chloride
Molecular Weight (g/mol): 219.46
Molecular Formula: C7H4BrClO
InChi Key: NZCKTGCKFJDGFD-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CC=C1Br
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results