accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH(R)(S)H2N

CAS RN 71581-92-5

IUPAC Name: (1S,2R)-N-benzyl-2-(hydroxymethyl)cyclohexan-1-aminium
Synonyms: (1S,2R)-N-benzyl-2-(hydroxymethyl)cyclohexan-1-aminium
Molecular Weight (g/mol): 220.34
Molecular Formula: C14H22NO
InChi Key: BRQFIORUNWWNBM-KBPBESRZSA-O
SMILES: OC[C@@H]1CCCC[C@@H]1[NH2+]CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results