accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 716-03-0

H3CNFOOH

CAS RN 716-03-0

IUPAC Name: 6-fluoro-2-methylquinoline-4-carboxylic acid
Synonyms: 6-fluoro-2-methylquinoline-4-carboxylic acid
Molecular Weight (g/mol): 205.19
Molecular Formula: C11H8FNO2
InChi Key: KXMSETXWKPJCQS-UHFFFAOYSA-N
SMILES: CC1=CC(C(O)=O)=C2C=C(F)C=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results